Learn More
N-des-succinyl-N-Boc-O-desethyl Sacubitril, TRC
Product Code. 30338202
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30338202 Supplier Toronto Research Chemicals Supplier No. TRCB4129651G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic Acid
CAS 1012341-50-2
Molecular Formula C23H29NO4
Quantity 1 g
Synonym (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic Acid, (alphaR,gammaS)-gamma-[[(1,1-Dimethylethoxy)carbonyl]amino]-alpha-methyl[1,1'-biphenyl]-4-pentanoic Acid
InChI Formula InChI=1S/C23H29NO4/c1-16(21(25)26)14-20(24-22(27)28-23(2,3)4)15-17-10-12-19(13-11-17)18-8-6-5-7-9-18/h5-13,16,20H,14-15H2,1-4H3,(H,24,27)(H,25,26)/t16-,20+/m1/s1
SMILES C[C@H](C[C@@H](Cc1ccc(cc1)c2ccccc2)NC(=O)OC(C)(C)C)C(=O)O
IUPAC Name (2R,4S)-2-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(4-phenylphenyl)pentanoic acid
Molecular Weight (g/mol) 383.48
Formula Weight 383.2097