missing translation for 'onlineSavingsMsg'
Learn More

N-Deshydroxyethyl Dasatinib, TRC

Product Code. 30132515
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132515 25 mg 25mg
30135788 5 mg 5mg
17970535 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132515 Supplier Toronto Research Chemicals Supplier No. TRCD29000025MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 910297-51-7
Molecular Formula C20 H22 Cl N7 O S
Molecular Weight (g/mol) 443.95
IUPAC Name N-(2-chloro-6-methylphenyl)-2-[(2-methyl-6-piperazin-1-ylpyrimidin-4-yl)amino]-1,3-thiazole-5-carboxamide
SMILES Cc1cccc(Cl)c1NC(=O)c2cnc(Nc3cc(nc(C)n3)N4CCNCC4)s2

Specifications

Quantity 25 mg
Synonym N-Deshydroxyethyl Dasatinib, 5-Thiazolecarboxamide, N-(2-chloro-6-methylphenyl)-2-[[2-methyl-6-(1-piperazinyl)-4-pyrimidinyl]amino]-, N-(2-Chloro-6-methylphenyl)-2-[[2-methyl-6-(1-piperazinyl)-4-pyrimidinyl]amino]-5-thiazolecarboxamide, N-Deshydroxyethyl dasatinib
InChI Formula InChI=1S/C20H22ClN7OS/c1-12-4-3-5-14(21)18(12)27-19(29)15-11-23-20(30-15)26-16-10-17(25-13(2)24-16)28-8-6-22-7-9-28/h3-5,10-11,22H,6-9H2,1-2H3,(H,27,29)(H,23,24,25,26)
Formula Weight 443.1295
Percent Purity >95%
Chemical Name or Material N-Deshydroxyethyl Dasatinib
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.