missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Ibandronate Sodium, TRC

Product Code. 30365085
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365085 10 mg 10mg
30352864 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365085 Supplier Toronto Research Chemicals Supplier No. TRCD29175510MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953805-81-7
Molecular Formula C8 H20 N O7 P2 . Na
Molecular Weight (g/mol) 327.18
Synonym Phosphonic acid, P,P'-[1-hydroxy-3-(pentylamino)propylidene]bis-, sodium salt (1:1), N-Desmethylibandronate Sodium, [1-Hydroxy-3-(pentylamino)propane-1,1-diyl]bis(phosphonic acid) Sodium, Ibandronate Sodium Monohydrate Imp. C (EP) als sodium salt
IUPAC Name sodium;hydroxy-[1-hydroxy-3-(pentylamino)-1-phosphonopropyl]phosphinate
SMILES [Na+].CCCCCNCCC(O)(P(=O)(O)O)P(=O)(O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H21NO7P2.Na/c1-2-3-4-6-9-7-5-8(10,17(11,12)13)18(14,15)16;/h9-10H,2-7H2,1H3,(H2,11,12,13)(H2,14,15,16);/q;+1/p-1
Formula Weight 327.0613
Percent Purity >95%
Chemical Name or Material N-Desmethyl Ibandronate sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.