missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Itopride Hydrochloride, TRC

Product Code. 30132978
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132978 25 mg 25mg
17914265 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132978 Supplier Toronto Research Chemicals Supplier No. TRCD29239025MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5922-37-2
Molecular Formula C19 H24 N2 O4 . Cl H
Molecular Weight (g/mol) 380.86
Synonym 3,4-dimethoxy-N-(4-(2-(methylamino)ethoxy)benzyl)benzamide, hydrochloride (1:1)
IUPAC Name 3,4-dimethoxy-N-[[4-[2-(methylamino)ethoxy]phenyl]methyl]benzamide;hydrochloride
SMILES Cl.CNCCOc1ccc(CNC(=O)c2ccc(OC)c(OC)c2)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H24N2O4.ClH/c1-20-10-11-25-16-7-4-14(5-8-16)13-21-19(22)15-6-9-17(23-2)18(12-15)24-3;/h4-9,12,20H,10-11,13H2,1-3H3,(H,21,22);1H
Formula Weight 380.1503
Percent Purity >95%
Chemical Name or Material Itopride Impurity 12 HCl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.