Learn More
N-Desmethyl-N-benzyl Sildenafil, TRC
Product Code. 30132690
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132690 500 mg 500mg
17922955 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30132690 Supplier Toronto Research Chemicals Supplier No. TRCD292205500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1446089-82-2
Molecular Formula C28 H34 N6 O4 S
Molecular Weight (g/mol) 550.67
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[[4-(phenylmethyl)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 5-[2-Ethoxy-5-[[4-(phenylmethyl)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Benzyl Sildenafil, Sildenafil-N-benzyl
IUPAC Name 5-[5-(4-benzylpiperazin-1-yl)sulfonyl-2-ethoxyphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(Cc5ccccc5)CC4
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C28H34N6O4S/c1-4-9-23-25-26(32(3)31-23)28(35)30-27(29-25)22-18-21(12-13-24(22)38-5-2)39(36,37)34-16-14-33(15-17-34)19-20-10-7-6-8-11-20/h6-8,10-13,18H,4-5,9,14-17,19H2,1-3H3,(H,29,30,35)
Formula Weight 550.2362
Percent Purity >95%
Chemical Name or Material N-Desmethyl-N-benzyl Sildenafil
Grade HPLC