missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl-N-hydroxymethyl Tebuthiuron, TRC

Product Code. 30180103
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180103 5 mg 5mg
30180063 25 mg 25mg
30180142 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180103 Supplier Toronto Research Chemicals Supplier No. TRCD2954455MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59962-54-8
Molecular Formula C9 H16 N4 O2 S
Molecular Weight (g/mol) 244.31
Synonym Urea, N-[5-(1,1-dimethylethyl)-1,3,4-thiadiazol-2-yl]-N'-(hydroxymethyl)-N-methyl, N-[5-(1,1-Dimethylethyl)-1,3,4-thiadiazol-2-yl]-N'-(hydroxymethyl)-N-methylurea
IUPAC Name 1-(5-tert-butyl-1,3,4-thiadiazol-2-yl)-3-(hydroxymethyl)-1-methylurea
SMILES CN(C(=O)NCO)c1nnc(s1)C(C)(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H16N4O2S/c1-9(2,3)6-11-12-8(16-6)13(4)7(15)10-5-14/h14H,5H2,1-4H3,(H,10,15)
Formula Weight 244.0994
Chemical Name or Material Tebuthiuron-N-hydroxymethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.