missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Nicergoline, TRC

Product Code. 30133428
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133428 25 mg 25mg
30136076 50 mg 50mg
17991695 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133428 Supplier Toronto Research Chemicals Supplier No. TRCD42236025MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192504-81-7
Molecular Formula C23 H24 Br N3 O3
Molecular Weight (g/mol) 470.36
Synonym Ergoline-8-methanol, 10-methoxy-1-methyl-, 5-bromo-3-pyridinecarboxylate (ester), (8β)- (9CI), N6-Demethylnicergoline, [(6aR,9R,10aS)-10a-Methoxy-4-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methyl 5-bromopyridine-3-carboxylate, 6-Demethylnicergoline, 6-Desmethylnicergoline, Nicergoline Imp. H (EP)
IUPAC Name [(6aR,9R,10aS)-10a-methoxy-4-methyl-6,6a,7,8,9,10-hexahydroindolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
SMILES CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN[C@@H]1Cc4cn(C)c5cccc2c45

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H24BrN3O3/c1-27-12-16-7-20-23(29-2,18-4-3-5-19(27)21(16)18)8-14(9-26-20)13-30-22(28)15-6-17(24)11-25-10-15/h3-6,10-12,14,20,26H,7-9,13H2,1-2H3/t14-,20-,23+/m1/s1
Formula Weight 469.1001
Percent Purity >95%
Chemical Name or Material 6-Demethylnicergoline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.