missing translation for 'onlineSavingsMsg'
Learn More

N-Despropyl Pergolide, TRC

Product Code. 30368569
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368569 2 mg 2mg
30361946 10 mg 10mg
30353240 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368569 Supplier Toronto Research Chemicals Supplier No. TRCD2973102MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72821-91-1
Molecular Formula C16 H20 N2 S
Molecular Weight (g/mol) 272.41
IUPAC Name (6aR,9R,10aR)-9-(methylsulfanylmethyl)-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline
SMILES CSC[C@H]1CN[C@@H]2Cc3c[nH]c4cccc([C@H]2C1)c34

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C16H20N2S/c1-19-9-10-5-13-12-3-2-4-14-16(12)11(8-18-14)6-15(13)17-7-10/h2-4,8,10,13,15,17-18H,5-7,9H2,1H3/t10-,13-,15-/m1/s1
Formula Weight 272.1347
Chemical Name or Material N-Despropyl Pergolide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.