missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyl-d25-amine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 1-Dodecylamine-D25 |
| CAS | 1219802-72-8 |
| Molecular Formula | C12H2D25N |
| Quantity | 0.01 g |
| Synonym | 1-Dodecanamine-d25, Dodecylamine-d25, 1-Aminododecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25, 1-Dodecylamine-D25, Laurylamine-d25, n-Dodecylamine-d25 |
| InChI Formula | InChI=1 S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | C(C(C(C(C(C(C([2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])(C(C(C(C(C(N)([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H] |
| IUPAC Name | dodecan-d25-1-amine |
| Molecular Weight (g/mol) | 210.51 |
| Formula Weight | 210.37 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction