missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyltriethoxysilane, 95%

Product Code. 11368376
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368376 25 g 25g
11378376 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368376 Supplier Thermo Scientific Alfa Aesar Supplier No. L17293.14
  • ONLINE EXCLUSIVE PRICE
    96.80€
    87.65€ / 25g
    Save 9.15€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18536-91-9 | C18H40O3Si | 332.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18536-91-9
Molecular Formula C18H40O3Si
Molecular Weight (g/mol) 332.6
MDL Number MFCD00048437
InChI Key YGUFXEJWPRRAEK-UHFFFAOYSA-N
Synonym silane, dodecyltriethoxy, n-dodecyltriethoxysilane, lauryl triethoxy silane, dodecyl triethoxy silane, acmc-209emw, silane,dodecyltriethoxy, dodecyltriethoxysilane, technical
PubChem CID 87696
IUPAC Name dodecyl(triethoxy)silane
SMILES CCCCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 152°C to 153°C (3mmHg)
Quantity 25 g
Linear Formula CH3(CH2)11Si(OCH2CH3)3
Beilstein 2414223
Flash Point >110°C (230°F)
Formula Weight 332.6
Percent Purity 95%
Refractive Index 1.433
Sensitivity Moisture sensitive
Density 0.89
Chemical Name or Material n-Dodecyltriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.