missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-2-pyrrolidone, Dr. Ehrenstorfer

Product Code. 30312084
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312084 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312084 Supplier Dr. Ehrenstorfer Supplier No. DRECA13354000
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethyl-2-pyrrolidone
CAS 2687-91-4
Analyte or Component Names N-Ethyl-2-pyrrolidone
Shipping Condition +20°C
Molecular Formula C6 H11 N O
Certifications/Compliance ISO 17034
Synonym 1-Ethyl-2-pyrrolidinone, 1-Ethyl-2-pyrrolidone, 1-Ethylazacyclopentan-2-one, E 0358, N-Ethyl-2-pyrrolidinone, N-Ethyl-2-pyrrolidone, N-Ethyl-α-pyrrolidinone, N-Ethyl-γ-butyrolactam, N-Ethylbutyrolactam, N-Ethylpyrrolidinone, N-Ethylpyrrolidone, NEP, NEP 100
InChI Formula InChI=1S/C6H11NO/c1-2-7-5-3-4-6(7)8/h2-5H2,1H3
SMILES CCN1CCCC1=O
IUPAC Name 1-ethylpyrrolidin-2-one
Molecular Weight (g/mol) 113.16
Quantity 250 mg
Formula Weight 113.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.