missing translation for 'onlineSavingsMsg'
Learn More

N-ethyl-3,4-dimethoxyaniline, TRC

Product Code. 30387760
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387760 100 mg
30383414 250 mg
30372705 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387760 Supplier Toronto Research Chemicals Supplier No. TRCE927650100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32953-13-2
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.23
Synonym N-Ethyl-3,4-dimethoxybenzenamine, 3,4-Dimethoxy-N-ethylaniline, N-(3,4-Dimethoxyphenyl)-N-ethylamine, N-(3,4-Dimethoxyphenyl)ethylamine, N-Ethyl-3,4-dimethoxyaniline
SMILES CCNc1ccc(OC)c(OC)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H15NO2/c1-4-11-8-5-6-9(12-2)10(7-8)13-3/h5-7,11H,4H2,1-3H3
Formula Weight 181.1103
Chemical Name or Material N-ethyl-3,4-dimethoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.