missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide Phosphate Diester, TRC

Product Code. 30376301
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376301 250 mg
30384876 100 mg
30378287 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376301 Supplier Toronto Research Chemicals Supplier No. TRCE678835250MG

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2965-52-8
Molecular Formula C24 H19 F34 N2 O8 P S2
Molecular Weight (g/mol) 1204.47
Synonym 1-Octanesulfonamide, N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-, phosphate (ester) (2:1) (8CI), N,N'-[Phosphinicobis(oxy-2,1-ethanediyl)]bis[N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide], N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide phosphate diester, diSAmPAP, 1-Octanesulfonamide, N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-, phosphate (ester) (2:1) (8CI), N,N'-[Phosphinicobis(oxy-2,1-ethanediyl)]bis[N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide], N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide phosphate diester
IUPAC Name bis[2-[ethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonyl)amino]ethyl] hydrogen phosphate
SMILES CCN(CCOP(=O)(O)OCCN(CC)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C24H19F34N2O8PS2/c1-3-59(70(63,64)23(55,56)19(45,46)15(37,38)11(29,30)9(25,26)13(33,34)17(41,42)21(49,50)51)5-7-67-69(61,62)68-8-6-60(4-2)71(65,66)24(57,58)20(47,48)16(39,40)12(31,32)10(27,28)14(35,36)18(43,44)22(52,53)54/h3-8H2,1-2H3,(H,61,62)
Formula Weight 1203.9778
Chemical Name or Material Bis(2-perfluorooctylsulfonyl-N-ethylaminoethyl) phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.