missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-nitrosomethallylamine, TRC

Product Code. 30178985
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178985 25 mg 25mg
30179591 100 mg 100mg
30178925 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30178985 Supplier Toronto Research Chemicals Supplier No. TRCE35773525MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68630-39-7
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
Synonym N-Nitroso-N-ethyl-N-(2-methylallyl)amine, N-Ethyl-2-methyl-N-nitroso-2-propen-1-amine
IUPAC Name N-ethyl-N-(2-methylprop-2-enyl)nitrous amide
SMILES CCN(CC(=C)C)N=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H12N2O/c1-4-8(7-9)5-6(2)3/h2,4-5H2,1,3H3
Formula Weight 128.095
Chemical Name or Material N-Ethyl-N-nitrosomethallylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.