missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(perfluoro-1-octanesulfonyl) Glycine, TRC

Product Code. 18073440
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073440 50 mg 50mg
18023350 5 mg 5mg
18003150 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18073440 Supplier Toronto Research Chemicals Supplier No. TRCE92551850MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2991-50-6
Molecular Formula C12 H8 F17 N O4 S
Molecular Weight (g/mol) 585.23
Synonym Glycine, N-ethyl-N-[(heptadecafluorooctyl)sulfonyl]- (7CI,8CI,9CI), N-Ethyl-N-[(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)sulfonyl]glycine, 2-(N-Ethylperfluorooctanesulfoamido)acetic acid, 2-(N-Ethylperfluorooctanesulfonamido)acetic acid, Et-PFOSA-AcOH, N-Ethyl-N-heptadecylfluorooctane sulfonyl glycine, N-Ethylperfluorooctanesulfonamidoacetate, N-EtFOSAA, EtFOSAA
IUPAC Name 2-[ethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonyl)amino]acetic acid
SMILES CCN(CC(=O)O)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H8F17NO4S/c1-2-30(3-4(31)32)35(33,34)12(28,29)10(23,24)8(19,20)6(15,16)5(13,14)7(17,18)9(21,22)11(25,26)27/h2-3H2,1H3,(H,31,32)
Formula Weight 584.9903
Percent Purity >95%
Chemical Name or Material 2-(N-Ethylperfluorooctanesulfonamido)acetic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.