missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethylamino) Fluvoxamine (up to 15% Z isomer), TRC

Product Code. 17932585
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932585 1 mg 1mg
17983735 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17932585 Supplier Toronto Research Chemicals Supplier No. TRCE8982801MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217262-11-7
Molecular Formula C17 H26 F3 N3 O2
Molecular Weight (g/mol) 361.4
Synonym 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-[2-[(2-aminoethyl)amino]ethyl]oxime, N-[2-[[[5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethyl]ethane-1,2-diamine, N-(Ethylamino) Fluvoxamine, N-(Ethylamino)fluvoxamine, EP Impurity F, 5-Methoxy-1-[4-(trifluoromethyl)phenyl]- 1-pentanone O-[2-[(2-Aminoethyl)amino]ethyl]oxime, N-[2-[[[5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethyl]ethane-1,2-diamine, N-(Ethylamino) Fluvoxamine (up to 15% Z isomer)
IUPAC Name N'-[2-[[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethyl]ethane-1,2-diamine
SMILES COCCCCC(=NOCCNCCN)c1ccc(cc1)C(F)(F)F

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H26F3N3O2/c1-24-12-3-2-4-16(23-25-13-11-22-10-9-21)14-5-7-15(8-6-14)17(18,19)20/h5-8,22H,2-4,9-13,21H2,1H3
Formula Weight 361.1977
Chemical Name or Material N-(Ethylamino) Fluvoxamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.