missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-alanine, 95%

Product Code. 15478908
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478908 5 g 5g
15488908 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478908 Supplier Thermo Scientific Alfa Aesar Supplier No. H62395.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 79990-15-1 | C18H17NO4 | 311.34 g/mol

N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.

Solubility
Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.

Notes
Refrigerate at 4°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79990-15-1
Molecular Formula C18H17NO4
Molecular Weight (g/mol) 311.34
MDL Number MFCD00062960
InChI Key QWXZOFZKSQXPDC-UHFFFAOYNA-N
Synonym fmoc-d-ala-oh, fmoc-d-alanine, n-fmoc-d-alanine, n-9-fluorenylmethoxycarbonyl-d-alanine, d-alanine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, n-9h-fluoren-9ylmethoxy carbonyl-d-alanine, r-2-9h-fluoren-9-yl-methoxycarbonylamino-propionic acid
PubChem CID 2724627
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 151°C to 157°C
Quantity 5 g
Beilstein 8025133
Solubility Information Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.
Formula Weight 311.34
Percent Purity 95%
Chemical Name or Material N-Fmoc-D-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.