missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-tyrosine Benzyl Ester, TRC

Product Code. 30398205
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398205 50 mg
30396007 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398205 Supplier Toronto Research Chemicals Supplier No. TRCF67972050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82911-77-1
Molecular Formula C31H27NO5
Molecular Weight (g/mol) 493.55
Synonym N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine Phenylmethyl Ester, (S)-Benzyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxyphenyl)propanoate
SMILES Oc1ccc(C[C@H](NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)OCc5ccccc5)cc1

Specifications

InChI Formula InChI=1S/C31H27NO5/c33-23-16-14-21(15-17-23)18-29(30(34)36-19-22-8-2-1-3-9-22)32-31(35)37-20-28-26-12-6-4-10-24(26)25-11-5-7-13-27(25)28/h1-17,28-29,33H,18-20H2,(H,32,35)/t29-/m0/s1
Quantity 50 mg
Formula Weight 493.1889
Shipping Condition Room Temperature
Chemical Name or Material N-Fmoc-L-tyrosine Benzyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.