missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Norfloxacin, TRC

Product Code. 17970925
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970925 10 mg 10mg
17951745 25 mg 25mg
17952295 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970925 Supplier Toronto Research Chemicals Supplier No. TRCF70101510MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70459-04-0
Molecular Formula C17 H18 F N3 O4
Molecular Weight (g/mol) 347.34
Synonym 1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Norfloxacin Imp. G (EP)
IUPAC Name 1-ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(CC3)C=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H18FN3O4/c1-2-20-9-12(17(24)25)16(23)11-7-13(18)15(8-14(11)20)21-5-3-19(10-22)4-6-21/h7-10H,2-6H2,1H3,(H,24,25)
Formula Weight 347.1281
Chemical Name or Material 1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.