missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, MP Biomedicals™

Product Code. 11428312 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.5 mL
100 mL
250 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428312 250 mL 250mL
11471142 0.5 mL 500mL
11461142 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428312 Supplier MP Biomedicals Supplier No. 0219522083
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

n-Hexane, ≥99%, Liquid

  • Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP. Hexadecane (n-Hexadecane) is an aliphatic long-chain hydrocarbon. It is a referen
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point °C to 95°C (literature)
Color 1 (APHA)
Density 0.66 at 20°C (water = 1); 0.659g/mL at 25°C(Lit.)
Quantity 250 mL
Flash Point -26°C / -14.8°F(Lit.)
Physical Form Liquid
Refractive Index n20/D 1.375(Lit.)
Residue after Evaporation <0 ppm
Specific Gravity 0.674
Vapor Pressure 124 mm Hg at 20°C 150 mm Hg at 24.8°C; 256 mmHg ( 37.7°C), 5.2 psi ( 37.7°C), ∽132 mmHg ( 20°C)(Lit.)
UN Number UN1208
Water <0.01%
Beilstein 1730733
Synonym Hexane, Esani, Gettysolve-B
Boiling Point 69°C (literature)
Solubility Information Soluble in water.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.