Learn More
N-Hydroxymethyl Sumatriptan, TRC
Product Code. 30133958
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133958 100 mg 100mg
30005246 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133958 Supplier Toronto Research Chemicals Supplier No. TRCH948775100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1797905-62-4
Molecular Formula C15 H23 N3 O3 S
Molecular Weight (g/mol) 325.43
Synonym [3-[2-(Dimethylamino)ethyl]-1-(hydroxymethyl)-1H-indol-5-yl]-N-methyl-methanesulfonamide, 1H-Indole-5-methanesulfonamide, 3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)-N-methyl-, Sumatriptan Succinate Imp. C (EP), Sumatriptan Succinate Rel. Comp. C (USP), 1H-Indole-5-methanesulfonamide, 3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)-N-methyl-, N-(hydroxymethyl) Sumatriptan (GR61089X)
IUPAC Name 1-[3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)indol-5-yl]-N-methylmethanesulfonamide
SMILES CNS(=O)(=O)Cc1ccc2c(c1)c(CCN(C)C)cn2CO
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H23N3O3S/c1-16-22(20,21)10-12-4-5-15-14(8-12)13(6-7-17(2)3)9-18(15)11-19/h4-5,8-9,16,19H,6-7,10-11H2,1-3H3
Formula Weight 325.146
Chemical Name or Material N-Hydroxymethyl Sumatriptan