Learn More
N-Isopropyl-4-((2-methylhydrazinyl)methyl)benzamide hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 366-70-1 |
| Molecular Formula | C12H20ClN3O |
| MDL Number | MFCD00072082 |
| Quantity | 250 mg |
| SMILES | O=C(NC(C)C)C1=CC=C(CNNC)C=C1.[H]Cl |
| Molecular Weight (g/mol) | 257.76 |
| Percent Purity | 98% |
| Chemical Name or Material | N-Isopropyl-4-((2-methylhydrazinyl)methyl)benzamide hydrochloride |