missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylcyclohexylamine, TRC

Product Code. 30401313
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401313 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401313 Supplier Toronto Research Chemicals Supplier No. TRCI8244555G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Isopropylcyclohexylamine
CAS 1195-42-2
Molecular Formula C9H19N
Quantity 5 g
Synonym Cyclohexylisopropylamine, Isopropylcyclohexylamine, N-(1-Methylethyl)cyclohexanamine, N-Cyclohexyl-N-isopropylamine, N-Cyclohexylisopropylamine, N-Isopropyl-N-cyclohexylamine, N-Isopropylcyclohexanamine, N- Isopropylcyclohexylamine, NSC 86132
InChI Formula InChI=1S/C9H19N/c1-8(2)10-9-6-4-3-5-7-9/h8-10H,3-7H2,1-2H3
SMILES CC(C)NC1CCCCC1
IUPAC Name N-propan-2-ylcyclohexanamine
Molecular Weight (g/mol) 141.25
Formula Weight 141.1517
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.