missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylthiourea, 97%, Thermo Scientific™

Product Code. 11323266
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323266 5 g 5g
11313266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323266 Supplier Thermo Scientific Alfa Aesar Supplier No. L12605.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Isopropylthiourea is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-76-2
Molecular Formula C4H10N2S
Molecular Weight (g/mol) 118.198
MDL Number MFCD00041375
InChI Key POXAIQSXNOEQGM-UHFFFAOYSA-N
Synonym n-isopropylthiourea, isopropylthiourea, isopropyl thiourea, urea, 1-isopropyl-2-thio, usaf d-6, 1-isopropyl-2-thiourea, isopropyl-thiourea, 1-isopropylthiourea, propan-2-yl thiourea, 1-iso-propyl-2-thiourea
PubChem CID 1711921
IUPAC Name propan-2-ylthiourea
SMILES CC(C)NC(=S)N

Specifications

Melting Point 166°C to 170°C
Odor Odorless
Quantity 5 g
Beilstein 1743052
Formula Weight 118.2
Percent Purity 97%
Chemical Name or Material N-Isopropylthiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.