missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methyl-2-(triphenylphosphoranylidene)acetamide, TRC

Product Code. 30474300
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474300 500 mg 500mg
30474900 100 mg 100mg
30478664 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474300 Supplier Toronto Research Chemicals Supplier No. TRCM337775500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129986-67-0
Molecular Formula C22H22NO2P
Molecular Weight (g/mol) 363.39
SMILES CON(C)C(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H22NO2P/c1-23(25-2)22(24)18-26(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21/h3-18H,1-2H3
Chemical Name or Material N-Methoxy-N-methyl-2-(triphenylphosphoranylidene)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.