missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine, 97%, Thermo Scientific™

Product Code. 10567674
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10567674 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10567674 Supplier Thermo Scientific Maybridge Supplier No. CC62146DA
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 898598-62-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H14F3N3
MDL Number MFCD09817515
Quantity 1 g
Synonym n-methyl-2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzylamine, methyl 2-2-methyl-5-trifluoromethyl pyrazol-3-yl phenyl methyl amine, n-methyl-1-2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl phenyl methanamine, n-methyl-1-2-2-methyl-5-trifluoromethyl-3-pyrazolyl phenyl methanamine, n-methyl-1-2-2-methyl-5-trifluoromethyl pyrazol-3-yl phenyl methanamine, benzenemethanamine,n-methyl-2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl
InChI Key OTPQTKMWNNGSEX-UHFFFAOYSA-N
SMILES CNCC1=CC=CC=C1C2=CC(=NN2C)C(F)(F)F
IUPAC Name N-methyl-1-[2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]phenyl]methanamine
Molecular Weight (g/mol) 269.271
PubChem CID 24229664
Formula Weight 269.27
Percent Purity 97%
Chemical Name or Material N-Methyl-2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.