missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(1-piperazinyl)nicotinamide, 99%, Thermo Scientific™

Product Code. 11419730
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419730 250 mg 250mg
11429730 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419730 Supplier Thermo Scientific Alfa Aesar Supplier No. H50968.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902837-15-4
Molecular Formula C11H16N4O
Molecular Weight (g/mol) 220.28
MDL Number MFCD08061094
InChI Key PKJNZUNKHVUUBC-UHFFFAOYSA-N
Synonym n-methyl-2-piperazin-1-yl-nicotinamide, n-methyl-2-1-piperazinyl nicotinamide, n-methyl-2-piperazin-1-yl pyridine-3-carboxamide, n-methyl-2-1-piperazino nicotinamide, n-methyl-2-piperazin-1-yl nicotinamide, n-methyl 2-piperazinyl 3-pyridyl carboxamide, n-methyl-2-piperazin-1-ylnicotinamide, n-methyl-2-1-piperazinyl pyridine-3-carboxamide
PubChem CID 28054516
IUPAC Name N-methyl-2-(piperazin-1-yl)pyridine-3-carboxamide
SMILES CNC(=O)C1=C(N=CC=C1)N1CCNCC1

Specifications

Quantity 250 mg
Formula Weight 220.28
Percent Purity 99%
Chemical Name or Material N-Methyl-2-(1-piperazinyl)nicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.