missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2,4-dinitroaniline, Mikromol™

Product Code. 30457520
Change view
Click to view available options
Quantity:
25 mg
250 mg
4 x 25 mg
Unit Size:
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457520 4 x 25 mg -
30052146 250 mg 250mg
30051998 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457520 Supplier Mikromol Supplier No. MM0154.5000254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Methyl-2,4-dinitroaniline, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl-2,4-dinitroaniline
CAS 2044-88-4
Analyte or Component Names N-Methyl-2,4-dinitroaniline
Shipping Condition Room Temperature
Molecular Formula C7 H7 N3 O4
Certifications/Compliance ISO 17025
Synonym N'-Methyl-2,4-dinitroaniline, Benzenamine, N-methyl-2,4-dinitro-, Aniline, N-methyl-2,4-dinitro- (6CI,7CI,8CI), (2,4-Dinitrophenyl)methylamine, 1-(Methylamino)-2,4-dinitrobenzene, 2,4-Dinitro-N-methylaniline, N-Methyl-2,4-dinitroaniline, N-Methyl-2,4-dinitrobenzenamine, NSC 36958
InChI Formula InChI=1S/C7H7N3O4/c1-8-6-3-2-5(9(11)12)4-7(6)10(13)14/h2-4,8H,1H3
Solution Type Neat
SMILES CNc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name N-methyl-2,4-dinitroaniline
Molecular Weight (g/mol) 197.15
Quantity 4 x 25 mg
Formula Weight 197.0437 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.