missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-phenylethylamine, TRC

Product Code. 30136057
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136057 10 g 10g
30133281 5 g 5g
30006599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136057 Supplier Toronto Research Chemicals Supplier No. TRCM32195510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-08-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
Synonym N-Methylbenzeneethanamine, (2-Phenylethyl)methylamine, N-(Phenylethyl)methylamine, N-Methyl-2-phenethylamine, N-Methyl-2-phenylethan-1-amine, N-Methyl-2-phenylethanamine, N-Methyl-2-phenylethylamine, N-Methyl-N-(2-phenylethyl)amine, N-Methyl-N-phenethylamine, N-Methyl-beta-phenethylamine, N-Methyl-beta-phenylethylamine, N-Methylphenethylamine, N-Phenethyl-N-methylamine, N-Phenethylmethylamine, NSC 113957, [2-(Methylamino)ethyl]benzene
IUPAC Name N-methyl-2-phenylethanamine
SMILES CNCCc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H13N/c1-10-8-7-9-5-3-2-4-6-9/h2-6,10H,7-8H2,1H3
Formula Weight 135.1048
Chemical Name or Material N-Methyl-2-phenylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.