Learn More
N-Methyl-(4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine, 97%, Thermo Scientific™
Product Code. 10491464
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10491464 1 g Amber Glass Bottle 1g
10744755 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10491464 Supplier Thermo Scientific Maybridge Supplier No. CC18146DA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 1031843-28-3
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD11109322
InChI Key XCBYKJIAOCVNGY-UHFFFAOYSA-N
Synonym n-methyl-4-methyl-2-phenyl-1,3-oxazol-5-ylmethyl amine, methyl 4-methyl-2-phenyl-1,3-oxazol-5-yl methyl amine, 5-methylamino methyl-4-methyl-2-phenyl-1,3-oxazole, methyl 4-methyl-2-phenyl 1,3-oxazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methanamine, n-methyl-1-4-methyl-2-phenyl-1,3-oxazol-5-yl methylamine
PubChem CID 33589464
IUPAC Name N-methyl-1-(4-methyl-2-phenyl-1,3-oxazol-5-yl)methanamine
SMILES CC1=C(OC(=N1)C2=CC=CC=C2)CNC
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material N-Methyl-(4-methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine