missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Gatifloxacin, TRC

Product Code. 30469529
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469529 25 mg 25mg
30468605 10 mg 10mg
30464738 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469529 Supplier Toronto Research Chemicals Supplier No. TRCM30750025MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114213-69-3
Molecular Formula C20H24FN3O4
Molecular Weight (g/mol) 389.42
Synonym 1-Cyclopropyl-7-(3,4-dimethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic Acid
IUPAC Name 1-cyclopropyl-7-(3,4-dimethylpiperazin-1-yl)-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid
SMILES COc1c(N2CCN(C)C(C)C2)c(F)cc3C(=O)C(=CN(C4CC4)c13)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H24FN3O4/c1-11-9-23(7-6-22(11)2)17-15(21)8-13-16(19(17)28-3)24(12-4-5-12)10-14(18(13)25)20(26)27/h8,10-12H,4-7,9H2,1-3H3,(H,26,27)
Formula Weight 389.1751
Chemical Name or Material N-Methyl Gatifloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.