Learn More
N-Methyl-L-glutamic Acid, TRC
Product Code. 30012508
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012508 250 mg 250mg
30009456 100 mg 100mg
30004479 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012508 Supplier Toronto Research Chemicals Supplier No. TRCN240115250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6753-62-4
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
Synonym N-Methyl-L-glutamic Acid, (2S)-2-(Methylamino)pentanedioic Acid, N-Methylglutamic Acid;
SMILES CN[C@@H](CCC(=O)O)C(=O)O
Specifications
InChI Formula InChI=1S/C6H11NO4/c1-7-4(6(10)11)2-3-5(8)9/h4,7H,2-3H2,1H3,(H,8,9)(H,10,11)/t4-/m0/s1
Quantity 250 mg
Shipping Condition Room Temperature
Chemical Name or Material N-Methyl-L-glutamic Acid