missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-chloropropyl)homoveratrylamine Hydrochloride, TRC

Product Code. 30478808
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478808 100 mg 100mg
30465367 250 mg 250mg
30476867 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478808 Supplier Toronto Research Chemicals Supplier No. TRCM295155100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36770-75-9
Molecular Formula C14H22ClNO2 . HCl
Molecular Weight (g/mol) 308.2
Synonym N-(3-Chloropropyl)-3,4-dimethoxy-N-methylbenzeneethanamine Hydrochloride, SR 45813 Hydrochloride, Verapamil EP Impurity D (HCl)
IUPAC Name 3-chloro-N-[2-(3,4-dimethoxyphenyl)ethyl]-N-methylpropan-1-amine;hydrochloride
SMILES Cl.COc1ccc(CCN(C)CCCCl)cc1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H22ClNO2.ClH/c1-16(9-4-8-15)10-7-12-5-6-13(17-2)14(11-12)18-3;/h5-6,11H,4,7-10H2,1-3H3;1H
Formula Weight 307.1106
Chemical Name or Material N-Methyl-N-(3-chloropropyl)homoveratrylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.