missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N,N-bis(2-pyridylethyl)amine, TRC

Product Code. 30008650
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008650 1 g 1g
30008397 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008650 Supplier Toronto Research Chemicals Supplier No. TRCM2942151G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5452-87-9
Molecular Formula C15 H19 N3
Molecular Weight (g/mol) 241.33
Synonym 2-Pyridineethanamine, N-methyl-N-[2-(2-pyridinyl)ethyl]-, N-Methyl-N-[2-(2-pyridinyl)ethyl]-2-pyridineethanamine, Methylbis(2-pyridylethyl)amine, N-Methylbis[2-(2-pyridylethyl)]amine, N-Methyl-N,N-bis[2-(2-pyridylethyl)]amine, NSC 19005, N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine
IUPAC Name N-methyl-2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)ethanamine
SMILES CN(CCc1ccccn1)CCc2ccccn2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H19N3/c1-18(12-8-14-6-2-4-10-16-14)13-9-15-7-3-5-11-17-15/h2-7,10-11H,8-9,12-13H2,1H3
Formula Weight 241.1579
Percent Purity >95%
Chemical Name or Material N-Methyl-N,N-bis(2-pyridylethyl)amine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.