missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N,N-bis(2-pyridylethyl)amine Trihydrochloride, TRC

Product Code. 17955095
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17955095 250 mg 250mg
17904225 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17955095 Supplier Toronto Research Chemicals Supplier No. TRCM295255250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2095467-43-7
Molecular Formula C15 H19 N3 . 3 Cl H
Molecular Weight (g/mol) 350.71
Synonym 2-Pyridineethanamine, N-methyl-N-[2-(2-pyridinyl)ethyl]-, hydrochloride (1:3), N-Methyl-N-[2-(2-pyridinyl)ethyl]-2-pyridineethanamine trihydrochloride, Methylbis(2-pyridylethyl)amine trihydrochloride, N-Methylbis[2-(2-pyridylethyl)]amine trihydrochloride, N-Methyl-N,N-bis[2-(2-pyridylethyl)]amine trihydrochloride, N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine trihydrochloride
IUPAC Name N-methyl-2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)ethanamine;trihydrochloride
SMILES Cl.Cl.Cl.CN(CCc1ccccn1)CCc2ccccn2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H19N3.3ClH/c1-18(12-8-14-6-2-4-10-16-14)13-9-15-7-3-5-11-17-15;;;/h2-7,10-11H,8-9,12-13H2,1H3;3*1H
Formula Weight 349.0879
Percent Purity >95%
Chemical Name or Material N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.