missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N’-nitrosopiperazine, TRC

Product Code. 18072150
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072150 250 mg 250mg
18003350 50 mg 50mg
18053650 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18072150 Supplier Toronto Research Chemicals Supplier No. TRCM325800250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16339-07-4
Molecular Formula C5 H11 N3 O
Molecular Weight (g/mol) 129.16
Synonym Piperazine, 1-methyl-4-nitroso-, 1-Methyl-4-nitrosopiperazine, 1-Nitroso-4-methylpiperazine, N-Nitroso-N'-methylpiperazine, NSC 523886
IUPAC Name 1-methyl-4-nitrosopiperazine
SMILES CN1CCN(CC1)N=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H11N3O/c1-7-2-4-8(6-9)5-3-7/h2-5H2,1H3
Formula Weight 129.0902
Percent Purity >95%
Chemical Name or Material N-Methyl-N'-nitrosopiperazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.