missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(o-aminophenyl)anthranilic Acid Methyl Ester, TRC

Product Code. 30473829
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473829 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473829 Supplier Toronto Research Chemicals Supplier No. TRCM28695510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl-N-(o-aminophenyl)anthranilic Acid Methyl Ester
CAS 1346601-40-8
Molecular Formula C15H16N2O2
Quantity 10 mg
Synonym 2-Methyl-2-(o-aminophenyl)amino Benzoic Acid Methyl Ester
InChI Formula InChI=1S/C15H16N2O2/c1-17(14-10-6-4-8-12(14)16)13-9-5-3-7-11(13)15(18)19-2/h3-10H,16H2,1-2H3
SMILES COC(=O)c1ccccc1N(C)c2ccccc2N
IUPAC Name methyl 2-(2-amino-N-methylanilino)benzoate
Molecular Weight (g/mol) 256.3
Formula Weight 256.1212
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.