missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals

Product Code. 10063473 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10063473

Brand: Thermo Scientific Acros 126722500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-59-9
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00002848
InChI Key CRVGTESFCCXCTH-UHFFFAOYSA-N
Synonym methyldiethanolamine, n-methyliminodiethanol, n-methyldiethanolamine, 2,2'-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, methyl diethanolamine, ethanol, 2,2'-methylimino bis, n-methyldiethanolimine, n-methylaminodiglycol, 2,2'-methylimino diethanol
PubChem CID 7767
SMILES CN(CCO)CCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -21°C
Color Colorless to Yellow
Density 1.0380 g/mL
Boiling Point 243°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (HOCH2CH2)2NCH3
Refractive Index 1.4675 to 1.4695
Quantity 250 g
Beilstein 04, 284
Specific Gravity 1.038
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with benzene,soluble in most organic solvents (alcohol,esters,aceton and chlorinated hydrocarbons),soluble in diethylether and aliphatic hydrocarbons
IUPAC Name 2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Viscosity 101 mPa.s (20°C)
Formula Weight 119.16
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N-Methyldiethanolamine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyldiethanolamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.