missing translation for 'onlineSavingsMsg'
Learn More

N-Methyltrimethylacetamide, 96%

Product Code. 11322786
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322786 5 g 5g
11332786 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322786 Supplier Thermo Scientific Alfa Aesar Supplier No. L08916.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6830-83-7 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6830-83-7
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00043643
InChI Key QMKKJBRRKIKWFK-UHFFFAOYSA-N
Synonym n-methylpivalamide, n-methyltrimethylacetamide, 2,2,n-trimethylpropanamide, n,2,2-trimethylpropionamide, n-methyl pivalamide, 2,2-dimethyl-n-methylpropanamide, bae, n-methyl-pivalamide, propanamide,n,2,2-trimethyl
PubChem CID 138844
IUPAC Name N,2,2-trimethylpropanamide
SMILES CC(C)(C)C(=O)NC

Specifications

Melting Point 89°C to 93°C
Boiling Point 105°C to 110°C (15 mmHg)
Linear Formula (CH3)3CCONHCH3
Quantity 5 g
Beilstein 1744142
Formula Weight 115.18
Percent Purity 96%
Chemical Name or Material N-Methyltrimethylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.