Learn More
N-Methyltrimethylacetamide, 96%
Product Code. 11332786
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332786 25 g 25g
11322786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11332786 Supplier Thermo Scientific Alfa Aesar Supplier No. L08916.14
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 6830-83-7 | C6H13NO | 115.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6830-83-7
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00043643
InChI Key QMKKJBRRKIKWFK-UHFFFAOYSA-N
Synonym n-methylpivalamide, n-methyltrimethylacetamide, 2,2,n-trimethylpropanamide, n,2,2-trimethylpropionamide, n-methyl pivalamide, 2,2-dimethyl-n-methylpropanamide, bae, n-methyl-pivalamide, propanamide,n,2,2-trimethyl
PubChem CID 138844
IUPAC Name N,2,2-trimethylpropanamide
SMILES CC(C)(C)C(=O)NC
Specifications
Melting Point 89°C to 93°C
Boiling Point 105°C to 110°C (15 mmHg)
Linear Formula (CH3)3CCONHCH3
Quantity 25 g
Beilstein 1744142
Formula Weight 115.18
Percent Purity 96%
Chemical Name or Material N-Methyltrimethylacetamide