missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Bis(1,4-benzodioxane-2-carbonyl)piperazine, TRC

Product Code. 30180372
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180372 25 mg 25mg
30179806 100 mg 100mg
30180323 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180372 Supplier Toronto Research Chemicals Supplier No. TRCB41050525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617677-53-9
Molecular Formula C22 H22 N2 O6
Molecular Weight (g/mol) 410.42
Synonym 1,4-Bis(2,3-dihydro-1,4-benzodioxin-2-ylcarbonyl)piperazine, Doxazosin Mesilate Imp. C (EP)
IUPAC Name [4-(2,3-dihydro-1,4-benzodioxine-3-carbonyl)piperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone
SMILES O=C(C1COc2ccccc2O1)N3CCN(CC3)C(=O)C4COc5ccccc5O4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H22N2O6/c25-21(19-13-27-15-5-1-3-7-17(15)29-19)23-9-11-24(12-10-23)22(26)20-14-28-16-6-2-4-8-18(16)30-20/h1-8,19-20H,9-14H2
Formula Weight 410.1478
Chemical Name or Material 1,4-Bis(2,3-dihydro-1,4-benzodioxin-2-ylcarbonyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.