missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)benzenesulfonamide, TRC

Product Code. 30134442
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134442 2.5 g 2.5g
30008492 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134442 Supplier Toronto Research Chemicals Supplier No. TRCB4189102.5G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58023-19-1
Molecular Formula C10 H13 Cl2 N O2 S
Molecular Weight (g/mol) 282.19
Synonym NSC 240403;
IUPAC Name N,N-bis(2-chloroethyl)benzenesulfonamide
SMILES ClCCN(CCCl)S(=O)(=O)c1ccccc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H13Cl2NO2S/c11-6-8-13(9-7-12)16(14,15)10-4-2-1-3-5-10/h1-5H,6-9H2
Formula Weight 281.0044
Percent Purity >95%
Chemical Name or Material N,N-Bis(2-chloroethyl)benzenesulfonamide
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.