missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Desethylene Levofloxacin Hydrochloride, TRC

Product Code. 17937914
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17937914 5 mg 5mg
17920085 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17937914 Supplier Toronto Research Chemicals Supplier No. TRCD2893505MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346603-62-0
Molecular Formula C16 H18 F N3 O4 . Cl H
Molecular Weight (g/mol) 371.79
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-10-[[2-(methylamino)ethyl]amino]-7-oxo-, hydrochloride (1:1), (3S)-, (S)-9-Fluoro-2,3-dihydro-3-methyl-10-[2-(methylamino)ethylamino]-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid hydrochloride, (3S)-9-Fluoro-3-methyl-10-[[2-(methylamino)ethyl]amino]-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid hydrochloride, N,N'-Desethylene Levofloxacin hydrochloride
IUPAC Name (2S)-7-fluoro-2-methyl-6-[2-(methylamino)ethylamino]-10-oxo-4-oxa-1-azatricyclo[7.3.1.05,13]trideca-5(13),6,8,11-tetraene-11-carboxylic acid;hydrochloride
SMILES Cl.CNCCNc1c(F)cc2C(=O)C(=CN3[C@@H](C)COc1c23)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H18FN3O4.ClH/c1-8-7-24-15-12(19-4-3-18-2)11(17)5-9-13(15)20(8)6-10(14(9)21)16(22)23;/h5-6,8,18-19H,3-4,7H2,1-2H3,(H,22,23);1H/t8-;/m0./s1
Formula Weight 371.1048
Chemical Name or Material (S)-9-Fluoro-2,3-dihydro-3-methyl-10-[2-(methylamino)ethylamino]-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.