missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di(2-hydroxypropyl)-p-toluidine (>85%), TRC

Product Code. 30004198
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004198 10 g 10g
30003327 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004198 Supplier Toronto Research Chemicals Supplier No. TRCD44666510G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38668-48-3
Molecular Formula C13 H21 N O2
Molecular Weight (g/mol) 223.31
Synonym 1,1'-[(4-Methylphenyl)imino]bis[2-propanol] (ACI), 2-Propanol, 1,1'-(p-tolylimino)di- (7CI), 1-[(2-Hydroxypropyl)(4-methylphenyl)amino]propan-2-ol, Diisopropanol-p-toluidine, N,N-Bis(2-hydroxypropyl)-4-methylaniline, N,N-Bis(2-hydroxypropyl)-p-toluidine, N,N-Di(2-hydroxypropyl)-p-toluidine, N,N-Diisopropanol-p-toluidine
IUPAC Name 1-[N-(2-hydroxypropyl)-4-methylanilino]propan-2-ol
SMILES CC(O)CN(CC(C)O)c1ccc(C)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H21NO2/c1-10-4-6-13(7-5-10)14(8-11(2)15)9-12(3)16/h4-7,11-12,15-16H,8-9H2,1-3H3
Formula Weight 223.1572
Chemical Name or Material N,N-Di(2-hydroxypropyl)-p-toluidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.