missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diallylthiourea, 96%, Thermo Scientific™

Product Code. 11322636
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322636 25 g 25g
11312636 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322636 Supplier Thermo Scientific Alfa Aesar Supplier No. L08648.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6601-20-3
Molecular Formula C7H12N2S
Molecular Weight (g/mol) 156.247
MDL Number MFCD00014945
InChI Key DZZWKUMHMSNBSG-UHFFFAOYSA-N
Synonym 1,3-diallylthiourea, 1,3-diallyl-2-thiourea, diallylthiourea, n,n'-diallylthiourea, bisallylthiocarbamide, n,n'-bis-allylthiourea, thiourea, n,n'-di-2-propenyl, thiourea, n,n'-di-2-propen-1-yl, 1,3-bis prop-2-en-1-yl thiourea, 1,3-bis prop-2-enyl thiourea
PubChem CID 3034186
IUPAC Name 1,3-bis(prop-2-enyl)thiourea
SMILES C=CCNC(=S)NCC=C

Specifications

Melting Point 42°C to 45°C
Odor Odorless
Quantity 25 g
Beilstein 1754800
Formula Weight 156.25
Percent Purity 96%
Chemical Name or Material N,N'-Diallylthiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.