missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethylenediamine (2-Diethylaminoethylamine), TRC

Product Code. 30041028
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30041028 Supplier Toronto Research Chemicals Supplier No. TRCD4436355G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-36-7
Molecular Formula C6 H16 N2
Quantity 5 g
Synonym N,N-Diethylethane-1,2-diamine, Tiapride Hydrochloride Imp. C (EP), Metoclopramide Imp. E (EP)
InChI Formula InChI=1S/C6H16N2/c1-3-8(4-2)6-5-7/h3-7H2,1-2H3
SMILES CCN(CC)CCN
IUPAC Name N',N'-diethylethane-1,2-diamine
Molecular Weight (g/mol) 116.2
Formula Weight 116.1313
Chemical Name or Material N,N-diethylethane-1,2-diamine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.