missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide, 99%

Product Code. 11311617
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311617 500 g 500g
11301617 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311617 Supplier Thermo Scientific Alfa Aesar Supplier No. L04503.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 617-84-5 | C5H11NO | 101.15 g/mol

N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

Solubility
Miscible with acetone, benzene, alcohol and ether.  

Notes
Incompatible with strong oxidizing agents. Avoid heat, flames and spark.                      
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

Density 0.91
Boiling Point 178°C
Flash Point 65°C (149°F)
Odor Amine-like
Refractive Index 1.434
Quantity 500 g
Beilstein 1209392
Solubility Information Miscible with acetone,benzene,alcohol and ether.
Formula Weight 101.15
Percent Purity 99%
Chemical Name or Material N,N-Diethylformamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.