missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,3-propanediamine, 99%

Product Code. 11342206
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342206 1000 mL 1 L
11332206 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342206 Supplier Thermo Scientific Alfa Aesar Supplier No. L03539.AU
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-55-7 | C5H14N2 | 102.181 g/mol

Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to methyl hydrazine. DMAPA is also used directly as a hardener in epoxy resins in the plastics industry, as a cross linking agent for cellulose fibers in the paper industry and as an anti-shriking agent for leather. It is used as an intermediate in the production of binding agents, ion-exchange materials, flocculating agents (water treatment), cosmetic agents, washing and cleaning agents (betaines), additive for petrol and other fuels, polyurethane fibers and lubricants, dyes, agrochemicals, agents used in the photographic and textile industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to methyl hydrazine. DMAPA is also used directly as a hardener in epoxy resins in the plastics industry, as a cross linking agent for cellulose fibers in the paper industry and as an anti-shriking agent for leather. It is used as an intermediate in the production of binding agents, ion-exchange materials, flocculating agents (water treatment), cosmetic agents, washing and cleaning agents (betaines), additive for petrol and other fuels, polyurethane fibers and lubricants, dyes, agrochemicals, agents used in the photographic and textile industries.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-55-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00008216
InChI Key IUNMPGNGSSIWFP-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine
PubChem CID 7993
IUPAC Name N',N'-dimethylpropane-1,3-diamine
SMILES CN(C)CCCN

Specifications

Melting Point -70°C
Density 0.818
Boiling Point 126°C to 128°C
Flash Point 32°C (89°F)
Odor Ammonia-like
Linear Formula (CH3)2N(CH2)3NH2
Refractive Index 1.435
Quantity 1000 mL
UN Number UN2734
Beilstein 605293
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-1,3-propanediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyl-1,3-propanediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N1,N1-dimethyl-1,3-Propanediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.