missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-vinylaniline, TRC

Product Code. 30384312
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384312 50 mg -
30374684 100 mg 100mg
30387719 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384312 Supplier Toronto Research Chemicals Supplier No. TRCD46487350MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-80-7
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
IUPAC Name 4-ethenyl-N,N-dimethylaniline
SMILES CN(C)c1ccc(C=C)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13N/c1-4-9-5-7-10(8-6-9)11(2)3/h4-8H,1H2,2-3H3
Formula Weight 147.1048
Chemical Name or Material N,N-Dimethyl-4-vinylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.