Learn More
N,N-Dimethyltryptamine Oxide, TRC
Product Code. 30005651
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005651 500 mg 500mg
17949454 50 mg 50mg
17969754 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005651 Supplier Toronto Research Chemicals Supplier No. TRCD471625500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 948-19-6
Molecular Formula C12 H16 N2 O
Molecular Weight (g/mol) 204.27
Synonym N,N-Dimethyltryptamine N-oxide, N,N-dimethyl N-oxide-1H-indole-3-ethanamine
IUPAC Name 2-(1H-indol-3-yl)-N,N-dimethyl-ethanamine oxide
SMILES CN(=O)(C)CCc1c[nH]c2ccccc12
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C12H16N2O/c1-14(2,15)8-7-10-9-13-12-6-4-3-5-11(10)12/h3-6,9,13H,7-8H2,1-2H3
Formula Weight 204.1263
Percent Purity >95%
Chemical Name or Material N,N-DMT N-oxide
Grade HPLC